Learn More
Lead(II) acetate basic, ACS reagent
CAS: 51404-69-4 | C4H10O8Pb3 | 807.69 g/mol
Supplier: Thermo Scientific Chemicals 453220010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Lead(II) acetate basic | |
| 1 kg | |
| XAOPZCQFBUCRKR-UHFFFAOYSA-L | |
| diacetyloxylead;lead;dihydrate | |
| 75087083 | |
| ≥33.0% (basic Pb as PbO) | |
| 1.5% max. (105°C, 2 h) |
| 51404-69-4 | |
| C4H10O8Pb3 | |
| CC(=O)O[Pb]OC(=O)C.O.O.[Pb] | |
| 807.69 | |
| 807.69 | |
| ACS Reagent |
Chemical Identifiers
| 51404-69-4 | |
| 807.69 | |
| 75087083 | |
| CC(=O)O[Pb]OC(=O)C.O.O.[Pb] |
| C4H10O8Pb3 | |
| XAOPZCQFBUCRKR-UHFFFAOYSA-L | |
| diacetyloxylead;lead;dihydrate |
Safety and Handling
GHS H Statement Harmful if swallowed. Harmful if inhaled. May damage the unborn child. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects.
GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Obtain special instructions before use. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Danger
EINECSNumber : 257-175-3
RUO – Research Use Only