Learn More
Lead(II) acetate, basic, ACS, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 036393A3
Description
Lead(II) acetate is used in the production of sweeteners, cosmetics, astringent, used in the cleaning and maintenance of stainless steel firearm suppressors (silencers) and compensators, and also used in making of slow matches.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lead(II) acetate | |
1335-32-6 | |
2 kg | |
MFCD00013041 | |
14,5419 | |
Soluble in water. | |
ACS |
Basic | |
Powder | |
Pb(OOCCH3)2·2Pb(OH)2 | |
UN1616 | |
Lead subacetate | |
807.71 | |
Air Sensitive |
Chemical Identifiers
1335-32-6 | |
MFCD00013041 | |
Lead subacetate |
Safety and Handling
GHS H Statement
H360-H351-H373
May damage fertility or the unborn child.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P260-P281-P308+P313-P501c
H351-H360Df-H373-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD ACETATE
EINECSNumber : 215-630-3
RTECSNumber : OF8750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only