Learn More
Lead(II) acetate, basic
CAS: 1335-32-6 | C4H6O4Pb·2Pb(OH)2
$141.99 - $492.79
Chemical Identifiers
| CAS | 1335-32-6 |
|---|---|
| Molecular Formula | C4H6O4Pb·2Pb(OH)2 |
| MDL Number | MFCD00013041 |
| Synonym | Lead subacetate |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1676336
|
Thermo Scientific Chemicals
A1676336 |
500 g |
Each for $141.99
|
|
|||||
|
AAA167630E
|
Thermo Scientific Chemicals
A167630E |
2500 g |
Each for $492.79
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1335-32-6 | |
| C4H6O4Pb·2Pb(OH)2 | |
| MFCD00013041 | |
| Lead subacetate |
Specifications
| 1335-32-6 | |
| Pb(OOCCH3)2·2Pb(OH)2 | |
| 500 g | |
| Air sensitive | |
| Lead subacetate | |
| Lead(II) acetate, basic |
| C4H6O4Pb·2Pb(OH)2 | |
| MFCD00013041 | |
| UN1616 | |
| 14,5419 | |
| 807.72 |
Safety and Handling
GHS H Statement
H360-H351-H373
May damage fertility or the unborn child.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P260-P281-P308+P313-P501c
H351-H360Df-H373-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD ACETATE
EINECSNumber : 215-630-3
RTECSNumber : OF8750000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only