missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) bromide, 98+%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 198855000

 View more versions of this product

Catalog No. AC198855000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

916.0°C
373.0°C
10031-22-8
Fine Crystalline Powder
98% min. (Argentometry)
PbBr2
15,182
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
dibromolead
24831
≥98%
0.1% max.
6.66
Lead(II) bromide
98+%
White
500 g
Br2Pb
MFCD00011156
15, 5458
Solubility in water: 5g/L (20 c). Other solubilities: insoluble in alcohol
[Br-].[Br-].[Pb++]
367.00
367.01
Extra Pure
Glass bottle
6.6600g/mL

Chemical Identifiers

10031-22-8
367.00
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
24831
Br2Pb
MFCD00011156
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
[Br-].[Br-].[Pb++]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child.
Suspected of damaging fertility.

GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for b

GHS Signal Word: Danger

EINECSNumber : 233-084-4

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only