missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) bromide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Lead(II) bromide, 99.999%, Br2Pb, CAS Number-10031-22-8, dibromoplumbane, acmc-1bq2x, lead 2+ bromide, ccris 4220, lead dibromide, lead bromide pbbr2, lead ii bromide, pbbr2, leadbromide, lead bromide, 25g, (metals basis), 916 deg.C, White, 6.66 g/mL

$112.66 - $366.07

Chemical Identifiers

CAS 10031-22-8
Molecular Formula Br2Pb
Molecular Weight (g/mol) 367.00
MDL Number MFCD00011156
InChI Key ZASWJUOMEGBQCQ-UHFFFAOYSA-L
Synonym lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
PubChem CID 24831
IUPAC Name dibromolead
SMILES [Br-].[Br-].[Pb++]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1072006
View Documents
Thermo Scientific Chemicals
01072006
5 g
Each for $112.66
 
AA1072014
View Documents
Thermo Scientific Chemicals
01072014
25 g
Each for $366.07
 
Description

Description

It is used in the field of antirust, pigment and photograph. The molten lead(II) bromide acts as an electrolyte. Lead(II) bromide provides a high concentration of lead(II) ions and bromide ions to carry the current during the electrolysis process. The rare-earth-doped alkali-lead bromide crystals (potassium lead bromide (or) KPB; rubidium lead bromide or RPB) emerge as promising new low-phonon-energy host materials for mid-IR applications and are useful for solid state lasers. Hybrid organic/lead halide perovskites are promising materials for solar cell fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10031-22-8
367.00
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
24831
[Br-].[Br-].[Pb++]
Br2Pb
MFCD00011156
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
dibromolead

Specifications

916°C
10031-22-8
Powder
Br2Pb
MFCD00011156
14,5403
Slightly soluble in water; Insoluble in alcohol; Soluble in ammonia,alkali,KBr and NaBr.
[Br-].[Br-].[Pb++]
367.00
367.01
Puratronic™
6.66 g/mL
(metals basis)
373°C
White
5 g
PbBr2
UN2291
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
dibromolead
24831
99.999%
Odorless
Lead(II) bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P312-P314-P330-P501c

H302+H332-H360Df-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.

EINECSNumber : 233-084-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only