missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) carbonate, ACS, Thermo Scientific Chemicals

Maximum level of impurities: Insoluble in dilute acetic acid 0.02%, Cl 0.002%, Nitrate and nitrite (as NO3) P.T. (limit about 0.005%), Ca 0.01%, K 0.02%, Na 0.05%, Cd 0.002%, Fe 0.005%, Zn 0.003%

Supplier:  Thermo Scientific Chemicals 04011936

 View more versions of this product

Catalog No. AA4011936


Explore available promotions

Description

Description

Lead(II) carbonate is used in polymerization catalysts. It serves as a curing agent for silicon resins, heat-resistant lubricants, lead-acid storage batteries, wire-reinforced hoses, thermistors and heat stabilizers for polyvinyl chloride polymers. Further, it is used in electrophotography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lead(II) carbonate
Powder
CO3Pb
UN2291
Soluble in acid and alkali. Insoluble in water,alcohol and ammonia.
[Pb++].[O-]C([O-])=O
267.20
267.2
Odorless
598-63-0
500 g
MFCD00064631
lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3
MFEVGQHCNVXMER-UHFFFAOYSA-L
λ²-lead(2+) carbonate
11727
ACS

Chemical Identifiers

598-63-0
267.20
MFEVGQHCNVXMER-UHFFFAOYSA-L
11727
CO3Pb
MFCD00064631
lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3
[Pb++].[O-]C([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332;
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H350-H360Df-H373

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 209-943-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only