missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) Chloride [for Perovskite precursor], TCI America™

Supplier:  TCI America L02921G

Catalog No. L02921G


Explore available promotions

Description

Description

Specifications

Specifications

Lead(II) Chloride [for Perovskite precursor]
501°C
951°C
MFCD00011157
2291
HWSZZLVAJGOAAY-UHFFFAOYSA-L
λ²-lead(2+) dichloride
24459
Crystalline Powder
7758-95-4
White
Cl2Pb
1 g
lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry
[Cl-].[Cl-].[Pb++]
278.10
278.10

Chemical Identifiers

7758-95-4
278.10
HWSZZLVAJGOAAY-UHFFFAOYSA-L
24459
[Cl-].[Cl-].[Pb++]
Cl2Pb
MFCD00011157
lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry
λ²-lead(2+) dichloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (1)-0252

RTECSNumber : OF9450000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions