missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) chromate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1067830

 View more versions of this product

Catalog No. AAA1067830


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lead (II) chromate
7758-97-6
CrO4Pb
MFCD00011158
14,5405
MOUPNEIJQCETIW-UHFFFAOYSA-N
dioxido(dioxo)chromium;lead(2+)
24460
323.19
Odorless
844°C
250 g
PbCrO4
UN3077
lead chromate, lead ii chromate, phoenicochroite, plumbous chromate, chrome yellow, chromate de plomb, lead chromate vi, lead chromate pbcro4, unii-aa3229aous
[O-][Cr](=O)(=O)[O-].[Pb+2]
323.192
CHEBI:86257
98%
6.3

Chemical Identifiers

7758-97-6
323.192
MOUPNEIJQCETIW-UHFFFAOYSA-N
24460
dioxido(dioxo)chromium;lead(2+)
CrO4Pb
MFCD00011158
lead chromate, lead ii chromate, phoenicochroite, plumbous chromate, chrome yellow, chromate de plomb, lead chromate vi, lead chromate pbcro4, unii-aa3229aous
CHEBI:86257
[O-][Cr](=O)(=O)[O-].[Pb+2]
Safety and Handling

Safety and Handling

GHS H Statement
H350-H360-H373-H201
May cause cancer.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Explosive
mass explosion hazard.

P201-P202-P260-P281-P308+P313-P501c

H350-H360Df-H373

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 231-846-0

RTECSNumber : GB2975000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only