missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) Iodide (99.99%, trace metals basis) 98.0+%, TCI America™

Supplier:  TCI America L02791KG

 View more versions of this product

Catalog No. L02791KG


Explore available promotions

Description

Description

Specifications

Specifications

954°C
412°C
Crystalline Powder
I2Pb
2291
RQQRAHKHDFPBMC-UHFFFAOYSA-L
λ²-lead(2+) diiodide
24931
≥98.0% (T)
Lead(II) Iodide (99.99%, trace metals basis) [for Perovskite precursor]
10101-63-0
1 kg
MFCD00011163
lead ii iodide, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, diiodo-, e2-plumbane
[I-].[I-].[Pb++]
461.00
461.01

Chemical Identifiers

10101-63-0
461.00
RQQRAHKHDFPBMC-UHFFFAOYSA-L
24931
[I-].[I-].[Pb++]
I2Pb
MFCD00011163
lead ii iodide, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, diiodo-, e2-plumbane
λ²-lead(2+) diiodide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (1)-1139

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions