Learn More
Lead(II) nitrate, 99+%
CAS: 10099-74-8 | N2O6Pb | 331.20 g/mol
$144.28 - $262.27
Chemical Identifiers
| CAS | 10099-74-8 |
|---|---|
| Molecular Formula | N2O6Pb |
| Molecular Weight (g/mol) | 331.20 |
| MDL Number | MFCD00011153 |
| InChI Key | RLJMLMKIBZAXJO-UHFFFAOYSA-N |
| Synonym | lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt |
| PubChem CID | 24924 |
| ChEBI | CHEBI:37187 |
| SMILES | [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC211565000
|
Thermo Scientific Chemicals
211565000 |
500 g | Plastic bottle |
Each for $144.28
|
|
||||
|
AC211560010
|
Thermo Scientific Chemicals
211560010 |
1 kg | Plastic bottle |
Each for $262.27
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10099-74-8 | |
| 331.20 | |
| RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
| 24924 | |
| [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| N2O6Pb | |
| MFCD00011153 | |
| lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
| CHEBI:37187 |
Specifications
| 470.0°C | |
| White | |
| 500 g | |
| N2O6Pb | |
| MFCD00011153 | |
| lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
| RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
| λ²-lead(2+) dinitrate | |
| 24924 | |
| 331.2 | |
| Plastic bottle | |
| 4.5300g/mL |
| 10099-74-8 | |
| Crystalline Powder | |
| 99% min. (Complexometry) | |
| Pb(NO3)2 | |
| 15,5469 | |
| Solubility in water: 343g/L. | |
| [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 331.20 | |
| CHEBI:37187 | |
| ≥99% | |
| 4.53 | |
| Lead(II) nitrate |
Safety and Handling
GHS H Statement
Harmful if inhaled.
May damage the unborn child.
Suspected of damaging fertility.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume
GHS Signal Word: Danger
EINECSNumber : 233-245-9
RUO – Research Use Only