Learn More
Lead(II) nitrate, 99%, Thermo Scientific Chemicals
$60.52 - $392.23
Chemical Identifiers
CAS | 10099-74-8 |
---|---|
Molecular Formula | N2O6Pb |
Molecular Weight (g/mol) | 331.20 |
MDL Number | MFCD00011153 |
InChI Key | RLJMLMKIBZAXJO-UHFFFAOYSA-N |
Synonym | lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt |
PubChem CID | 24924 |
ChEBI | CHEBI:37187 |
IUPAC Name | lead(2+);dinitrate |
SMILES | [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1634522
|
Thermo Scientific Chemicals
A1634522 |
100 g |
Each for $60.52
|
|
|||||
AAA1634536
|
Thermo Scientific Chemicals
A1634536 |
500 g |
Each for $118.70
|
|
|||||
AAA163450E
|
Thermo Scientific Chemicals
A163450E |
2500 g |
Each for $392.23
|
|
|||||
Description
Lead(II) nitrate is employed in industrial applications such as heat stabilization in nylon and polyesters and coatings of photothermographic paper. It is utilized to improve the leaching process in the gold cyanidation. It finds applications as a lead paint as well as a source of dinitrogen tetroxide. As an oxidant, it is used in the oxidation of benzylic halides to aldehydes in organic synthesis. It is also used in the preparation of dithiocarbamates from isothiocyanates. It acts as a scavenger of bromide in nucleophilic substitution reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsLead(II) nitrate is employed in industrial applications such as heat stabilization in nylon and polyesters and coatings of photothermographic paper. It is utilized to improve the leaching process in the gold cyanidation. It finds applications as a lead paint as well as a source of dinitrogen tetroxide. As an oxidant, it is used in the oxidation of benzylic halides to aldehydes in organic synthesis. It is also used in the preparation of dithiocarbamates from isothiocyanates. It acts as a scavenger of bromide in nucleophilic substitution reactions.
Solubility
Soluble in water. Insoluble in nitric acid. Sparingly soluble in methanol.
Notes
Incompatible with strong reducing agents, organic materials and powdered metals.
Chemical Identifiers
10099-74-8 | |
331.20 | |
RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
24924 | |
lead(2+);dinitrate |
N2O6Pb | |
MFCD00011153 | |
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
CHEBI:37187 | |
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
470°C | |
100 g | |
MFCD00011153 | |
14,5414 | |
Soluble in water. Insoluble in nitric acid. Sparingly soluble in methanol. | |
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
331.20 | |
CHEBI:37187 | |
99% | |
4.53 |
10099-74-8 | |
N2O6Pb | |
UN1469 | |
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
lead(2+);dinitrate | |
24924 | |
331.2 | |
Odorless | |
Lead(II) nitrate |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P310-P330-P370+P378q-P501c
H272-H302+H332-H318-H336-H350-H360Df-H373
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: LEAD NITRATE
EINECSNumber : 233-245-9
RTECSNumber : OG2100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only