Learn More
Lead(II) nitrate, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 423855000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lead(II) nitrate | |
10099-74-8 | |
Crystalline Powder | |
N2O6Pb | |
MFCD00011153 | |
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
lead(2+);dinitrate | |
24924 | |
331.2 | |
ACS Reagent | |
4.53 |
470.0°C | |
White | |
500 g | |
Pb(NO3)2 | |
15, 5469 | |
Solubility in water: 343g/L. | |
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
331.20 | |
CHEBI:37187 | |
≥99.0% | |
Plastic bottle | |
4.5300g/mL |
Chemical Identifiers
10099-74-8 | |
331.20 | |
RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
24924 | |
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
N2O6Pb | |
MFCD00011153 | |
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
CHEBI:37187 |
Safety and Handling
GHS H Statement
Harmful if inhaled.
May damage the unborn child.
Suspected of damaging fertility.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume
GHS Signal Word: Danger
EINECSNumber : 233-245-9
RTECSNumber : OG2100000
TSCA : TSCA
RUO – Research Use Only