missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) Oxide, Extra Pure, MilliporeSigma™
Supplier: Technidata 1056585000
Specifications
| Lead(II) oxide | |
| Solid | |
| OPb | |
| UN 2291 | |
| YEXPOXQUZXUXJW-UHFFFAOYSA-N | |
| plumbanone | |
| 14827 | |
| Extra Pure | |
| Plastic bottle |
| 1317-36-8 | |
| 5 kg | |
| MFCD00011164 | |
| lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow | |
| O=[Pb] | |
| 223.20 | |
| 223.19 | |
| Odorless | |
| Yellow solid |
Chemical Identifiers
| 1317-36-8 | |
| 223.20 | |
| YEXPOXQUZXUXJW-UHFFFAOYSA-N | |
| 14827 | |
| O=[Pb] |
| OPb | |
| MFCD00011164 | |
| lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow | |
| plumbanone |
Safety and Handling
H360FD: May damage fertility. May damage the unborn child.
H302 + H332: Harmful if swallowed or if inhaled
H373: May cause damage to organs through prolonged or repeated exposure.
H410: Very toxic to aquatic life with long lasting effects.
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 215-267-0