missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) perchlorate trihydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 254590010

 View more versions of this product

Catalog No. AC254590010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

Lead(II) perchlorate trihydrate
99%
White to Beige
Adhering Crystals
98.5% min. (Complexometry)
Pb(ClO4)2·3H2O
lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent
KKGGEAQRICYXNM-UHFFFAOYSA-L
λ²-lead(2+) trihydrate diperchlorate
25021834
99%
2.6
100.0°C
13453-62-8
3 to 5 (5% soln.)
1 kg
Cl2H6O11Pb
MFCD00149817
Solubility in water: soluble. Other solubilities: soluble in alcohol
O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
460.10
460.14
Glass bottle
2.6000g/mL

Chemical Identifiers

13453-62-8
460.10
KKGGEAQRICYXNM-UHFFFAOYSA-L
25021834
Cl2H6O11Pb
MFCD00149817
lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent
O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
May damage the unborn child. Suspected of damaging fertility.
May intensify fire; oxidizer.

GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only