Learn More
Lead(II) sulfate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01073018
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lead(II) sulfate | |
7446-14-2 | |
50 g | |
O4PbS | |
UN2291 | |
lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite | |
[O-]S(=O)(=O)[O-].[Pb+2] | |
303.256 | |
303.25 | |
Puratronic™ | |
6.2 g/mL |
1170°C | |
Crystalline | |
(metals basis) | |
MFCD00011166 | |
14,5420 | |
KEQXNNJHMWSZHK-UHFFFAOYSA-L | |
lead(2+);sulfate | |
24008 | |
99.999% | |
Odorless |
Chemical Identifiers
7446-14-2 | |
303.256 | |
KEQXNNJHMWSZHK-UHFFFAOYSA-L | |
24008 | |
[O-]S(=O)(=O)[O-].[Pb+2] |
O4PbS | |
MFCD00011166 | |
lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite | |
lead(2+);sulfate |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H350-H360-H373
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 231-198-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only