missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) sulfate, Reagent Grade, Thermo Scientific Chemicals

For Use in Pigments, Galvanic Batteries, Lithography

Supplier:  Thermo Scientific Chemicals 04478422

 View more versions of this product

Catalog No. AA4478422


Only null left
Explore available promotions

Description

Description

Lead(II) sulfate is used in pigments, galvanic batteries, lithography, weighting fabrics. It is also used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lead(II) sulfate
7446-14-2
Lump/Powder
O4PbS
lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite
KEQXNNJHMWSZHK-UHFFFAOYSA-L
lead(2+);sulfate
24008
Reagent
White powder/lump
1170°C
White
100 g
MFCD00011166
Partially soluble in water,dilute HCl and HNO3. Soluble in sodium hydroxide,ammonium acetate or tartrate,concentrated hydriodic acid. Insoluble in alcohol
[O-]S(=O)(=O)[O-].[Pb+2]
303.256
303.25
Odorless
6.2 g/cm3 at 20°C

Chemical Identifiers

7446-14-2
303.256
KEQXNNJHMWSZHK-UHFFFAOYSA-L
24008
[O-]S(=O)(=O)[O-].[Pb+2]
O4PbS
MFCD00011166
lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite
lead(2+);sulfate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302+H312+H332-H350-H360-H373

DOTInformation : DOT Class: 9; Packaging Group: III

EINECSNumber : 231-198-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only