missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) sulfide, Pb 82% min., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1026836

 View more versions of this product

Catalog No. AAA1026836

Explore available promotions

Description

Description

Lead(II) sulfide is used as a semiconductor. Used in electronic devices and Infrared sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lead(II) sulfide is used as a semiconductor. Used in electronic devices and Infrared sensor.

Solubility
Soluble in water (0.00086g/L) and acid. Insoluble in alcohol, and potassium hydroxide.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Lead (II) sulfide
1314-87-0
PbS
UN3077
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
FILXVLQXZYXBSJ-UHFFFAOYSA-N
λ²-lead(2+) sulfanediide
14819
Pb 82% min.
3.921
1114°C
500 g
MFCD00016280
14,5421
Soluble in water (0.00086g/L) and acid. Insoluble in alcohol,and potassium hydroxide.
[S--].[Pb++]
239.30
239.25
Odorless
7.5

Chemical Identifiers

1314-87-0
239.30
FILXVLQXZYXBSJ-UHFFFAOYSA-N
14819
PbS
MFCD00016280
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
[S--].[Pb++]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H350-H360Df-H373

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 215-246-6

RTECSNumber : OG4550000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only