missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) titanium oxide, 99.5%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 1413922

 View more versions of this product

Catalog No. AA1413922


Explore available promotions

Description

Description

Lead(II) titanium oxide is widely used in ceramic capacitor, ceramic filters, PTC thermal resistor, pressure sensitive resistor and the performance of transducer field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lead(II) titanium oxide
Yellow
100 g
MFCD00011159
lead titanate, lead titanium trioxide, lead ii titanate, lead titanium oxide, lead titanium oxide pbtio3, titanic acid, lead salt, titanox l, titanicacid,leadsalt, o3ti.pb, lead titanate, powder,
NKZSPGSOXYXWQA-UHFFFAOYSA-N
dioxido(oxo)titanium;lead(2+)
16211560
99.5%
7.520 g/mL
12060-00-3
Powder
O3PbTi
UN3288
Insoluble in water.
[O-][Ti](=O)[O-].[Pb+2]
303.064
303.09
Odorless

Chemical Identifiers

12060-00-3
303.064
NKZSPGSOXYXWQA-UHFFFAOYSA-N
16211560
[O-][Ti](=O)[O-].[Pb+2]
O3PbTi
MFCD00011159
lead titanate, lead titanium trioxide, lead ii titanate, lead titanium oxide, lead titanium oxide pbtio3, titanic acid, lead salt, titanox l, titanicacid,leadsalt, o3ti.pb, lead titanate, powder,
dioxido(oxo)titanium;lead(2+)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H360-H373-H332
Fatal if swallowed.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P330-P501c

H300-H332-H350-H360Df-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 235-038-9

RTECSNumber : XR1575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only