missing translation for 'onlineSavingsMsg'
Learn More

Lead(IV) oxide, 97+%, extra pure

CAS: 1309-60-0 | O2Pb | 239.20 g/mol

$52.40 - $448.45

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.20
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
PubChem CID 14793
SMILES O=[Pb]=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC217530050
SDS View Documents
Thermo Scientific Chemicals
217530050
5 g Glass bottle
Each for $52.40
Only null left
 
AC217531000
SDS View Documents
Thermo Scientific Chemicals
217531000
100 g Glass bottle
Each for $161.96
Only null left
 
AC217535000
SDS View Documents
Thermo Scientific Chemicals
217535000
500 g Glass bottle
Each for $448.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

1309-60-0
239.20
YADSGOSSYOOKMP-UHFFFAOYSA-N
14793
O2Pb
MFCD00011165
lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
O=[Pb]=O

Specifications

290°C
Brown to Black
5 g
O2Pb
MFCD00011165
15, 5462
Solubility in water: insoluble. Other solubilities: soluble in hcl and glacial acetic acid,insoluble in alcohol
O=[Pb]=O
239.20
239.19
Extra Pure
Lead(IV) oxide, p.a.
1309-60-0
Powder
97+%
PbO2
01,5331; 02,233; 03,168; 11,282
lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
YADSGOSSYOOKMP-UHFFFAOYSA-N
dioxolead
14793
≥97%
Glass bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child.
Suspected of damaging fertility.

GHS P Statement
Use personal protective equipment as required.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON C

GHS Signal Word: Danger

EINECSNumber : 215-174-5

RTECSNumber : OG0700000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only