Learn More
Lead(IV) oxide, ACS, 97.0% min, Thermo Scientific Chemicals
For Use in Battery Electrodes, Manufacturing of Pigments and Dyes, As Analytical Lab Reagent
$124.66 - $563.51
Chemical Identifiers
CAS | 1309-60-0 |
---|---|
Molecular Formula | O2Pb |
Molecular Weight (g/mol) | 239.20 |
MDL Number | MFCD00011165 |
InChI Key | YADSGOSSYOOKMP-UHFFFAOYSA-N |
Synonym | lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french |
PubChem CID | 14793 |
SMILES | O=[Pb]=O |
Description
It finds applications as an electrode in lead acid batteries, as anode in electroplating copper and zinc in sulfate solution, production of glyoxylic acid, manufacturing of pigments and dyes, high voltage lightening arrestors, and as an analytical lab reagent. Lead dioxide is a potent oxidizing agent which is employed in the manufacture of matches, pyrotechnics and other chemicals. It can be used in the curing of sulfide-based polymers. It oxidizes Mn(II) into Mn(VII), and Cr(III) into Cr(VI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Maximum level of impurities: Acid-insoluble matter 0.2%, Carbon compounds (as C) 0.04%, Cl 0.002%, NO3 0.02%, SO4 0.05%, Mn 5ppm, Ca 0.02%, Cu 0.05%, K 0.05%, Na 0.1%Chemical Identifiers
1309-60-0 | |
239.20 | |
YADSGOSSYOOKMP-UHFFFAOYSA-N | |
14793 |
O2Pb | |
MFCD00011165 | |
lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french | |
O=[Pb]=O |
Specifications
290°C (decomposition) | |
Brown to Black | |
100 g | |
O2Pb | |
lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french | |
YADSGOSSYOOKMP-UHFFFAOYSA-N | |
plumbanedione | |
14793 | |
ACS | |
9.38 |
1309-60-0 | |
Powder | |
97.0% min. | |
MFCD00011165 | |
Insoluble in water and alcohol. Soluble in acetic acid. | |
O=[Pb]=O | |
239.20 | |
239.19 | |
Odorless | |
Lead(IV) oxide |
Safety and Handling
P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P370+P378q-P501c
H272-H302+H332-H350-H360Df-H373
DOTInformation : DOT Class: 5.1, Packaging Group: III
EINECSNumber : 215-174-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only