missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead Sulfate, Powder, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor L109512KG

 View more versions of this product

Catalog No. 18611260


Explore available promotions

Description

Description

Specifications

Specifications

7446-14-2
12 kg
Reagent
1
97%
Poly Pail

Chemical Identifiers

7446-14-2
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.