missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Sealed Overhead LED Light Fixture and Frame For Workstations
Sealed overhead LED light and frame are ideal for 60 to 72 in. benches.
Supplier: Fisherbrand™ LFA48+SLD24
Description
For 48, 60, or 72 in. Long Table, Instant Maximum Brightness, Cleanroom rated, Long lasting 12V LED light, creates less heat with less power consumption, Bayer polycrabonate housing for maximum difused light and long life, 5,000 Kelvin Color Temperature - similar to Fluorescent Daylight, Quiet and no-fluorescent-buzzing sound, Sealed to eliminate dust and reduce fire hazards, Stainless steel mounting brackets, No high voltage danger, 6A 115V power supply, 10 year manufacturer warranty
- For 48 in., 60 in., or 72 in. Long Tables
- Instant Maximum Brightness
- Cleanroom rated
- Long lasting 12V LED light, creates less heat with less power consumption
- Bayer polycarbonate housing for maximum difused light and long life
- 5,000 Kelvin Color Temperature - similar to Fluorescent Daylight
- Quiet and no-fluorescent-buzzing sound
- Sealed to eliminate dust and reduce fire hazards
- Stainless steel mounting brackets
- No high voltage danger
- 6A 115V power supply
- 10 year manufacturer warranty
Order Info
Must order upright post (Fisher SKU 25100350)
Warranty
25-year manufacturer warranty
Specifications
Light Fixture and Frame | |
12 V | |
6 A | |
48 in. light frame; 24 in. light fixture | |
4 ft. workstation |
LED | |
34 W | |
Polycarbonate, Stainless Steel | |
Workstation lighting | |
Switch and 6 A, 115 V power supply cord |