Supertek™ Lens, Double Convex DFS Item
Double Convex lens made of high quality glass with outstanding optical properties.
$5.82 - $5.82
Specifications
Product Type Double Convex Lens
Shape Convex Circle
View More Specs
Products 2
Catalog Number Mfr. No. Length (Metric) Focal Price Quantity  
Catalog Number Mfr. No. Length (Metric) Focal Price Quantity  
S012328 DFS Item
View Documents
Supertek™
PH300644
200 mm
Each for $5.82
S012327 DFS Item
View Documents
Supertek™
PH300646
300 mm
Each for $5.82
Description
Description
  • Double convex lens.
  • Made of high quality glass with outstanding optical properties.
  • 38 mm diameter, 200 or 300 mm Focal length
Specifications
Specifications
Double Convex Lens
Convex Circle
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'