missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leu-Enkephalin

CAS: 81678-16-2615.684 g/mol

Supplier:  Thermo Scientific Chemicals J61408MB

Catalog No. AAJ61408MB


Only null left
Explore available promotions

Description

Description

Endogenous opioid neurotransmitter/neuromodulator that is an agonist at ? and δ opioid receptors, localization in brain parallels the localization of δ receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Endogenous opioid neurotransmittereuromodulator that is an agonist at μ and δ opioid receptors, localization in brain parallels the localization of δ receptors.

Solubility
Soluble in water (1 mg/ml ).
Specifications

Specifications

81678-16-2
Store at -20°C
615.684
25 mg
White
Lyophilized Powder
Leu-Enkephalin

Chemical Identifiers

81678-16-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only