missing translation for 'onlineSavingsMsg'
Learn More

Leucoquinizarin 98.0+%, TCI America™

$87.60 - $1068.11

Chemical Identifiers

CAS 476-60-8
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
MDL Number MFCD00058948
InChI Key BKNBVEKCHVXGPH-UHFFFAOYSA-N
Synonym 2,3-Dihydro-9,10-dihydroxy-1,4-anthraquinone, 1,4-Dihydroxyanthrahydroquinone, leuco-Quinizarin, 1,4,9,10-Tetrahydroxyanthracene
PubChem CID 68063
IUPAC Name anthracene-1,4,9,10-tetrol
SMILES OC1=C2C(O)=C3C=CC=CC3=C(O)C2=C(O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T011625G
View Documents
TCI America
T011625G
25 g
Each for $87.60
Only null left
 
T0116500G
View Documents
TCI America
T0116500G
500 g
Each for $1,068.11
Only null left
 
Description

Description

Specifications

Chemical Identifiers

476-60-8
242.23
BKNBVEKCHVXGPH-UHFFFAOYSA-N
68063
OC1=C2C(O)=C3C=CC=CC3=C(O)C2=C(O)C=C1
C14H10O4
MFCD00058948
2,3-Dihydro-9,10-dihydroxy-1,4-anthraquinone, 1,4-Dihydroxyanthrahydroquinone, leuco-Quinizarin, 1,4,9,10-Tetrahydroxyanthracene
anthracene-1,4,9,10-tetrol

Specifications

476-60-8
C14H10O4
25 g
BKNBVEKCHVXGPH-UHFFFAOYSA-N
anthracene-1,4,9,10-tetrol
68063
≥98.0% (HPLC)
Leucoquinizarin
151°C
MFCD00058948
2,3-Dihydro-9,10-dihydroxy-1,4-anthraquinone, 1,4-Dihydroxyanthrahydroquinone, leuco-Quinizarin, 1,4,9,10-Tetrahydroxyanthracene
OC1=C2C(O)=C3C=CC=CC3=C(O)C2=C(O)C=C1
242.23
242.23
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : OH4742000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'