3M™ Lexa™ Replacement Lenses
Lexa protective eyewear replacement lens features an 3M™ DX™ Anti-Fog hard coat which helps protect lens from fog, scratching, and many chemicals. Polycarbonate lens absorbs 99.9% UV.
$95.71 - $118.64
Specifications
Lens Coating Anti-fog
Description Three lens angles, and four temple lengths
Product Type Replacement Lens
Lens Material Polycarbonate
View More Specs
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Products 2
Catalog Number Mfr. No. Lens Coating Lens Material Lens Tint Price Quantity  
Catalog Number Mfr. No. Lens Coating Lens Material Lens Tint Price Quantity  
19-034-451
Accessories View Documents
3M™
152450000020
Anti-fog Polycarbonate Clear
Pack of 20 Each for $118.64
19-034-452
View Documents
3M™
152460000020
Anti-fog Polycarbonate Gray
Pack of 20 Each for $95.71
Description
Description
Lexa protective eyewear replacement lens features an 3M™ DX™ Anti-Fog hard coat which helps protect lens from fog, scratching, and many chemicals. Polycarbonate lens absorbs 99.9% UV.
  • Gray, anti-fog lens
  • Large size
  • 20 safety glasses per case
Specifications
Specifications
Anti-fog
Three lens angles, and four temple lengths
Replacement Lens
Polycarbonate
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'