missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lidocaine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals

$71.96 - $193.58

Chemical Identifiers

CAS 6108-05-0
Molecular Formula C14H25ClN2O2
Molecular Weight (g/mol) 288.82
MDL Number MFCD00150329
InChI Key YECIFGHRMFEPJK-UHFFFAOYSA-N
Synonym lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate
PubChem CID 16219577
ChEBI CHEBI:60791
IUPAC Name {[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride
SMILES O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6303506
View Documents
Thermo Scientific Chemicals
J6303506
5 g
Each for $71.96
 
AAJ6303514
View Documents
Thermo Scientific Chemicals
J6303514
25 g
Each for $193.58
 
Description

Description

It is also used as a fast voltage-gated sodium channel blocker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is also used as a fast voltage-gated sodium channel blocker.

Solubility
Soluble in water (50 mg/ml), chloroform, ethanol, and benzene. Insoluble in diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Do not store above 40 degree C (104 0 F). (preferably store at temperature between 15degree C and 30 degree C).
Specifications

Chemical Identifiers

6108-05-0
288.82
YECIFGHRMFEPJK-UHFFFAOYSA-N
16219577
{[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride
C14H25ClN2O2
MFCD00150329
lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate
CHEBI:60791
O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C

Specifications

6108-05-0
White
MFCD00150329
UN2811
lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate
YECIFGHRMFEPJK-UHFFFAOYSA-N
{[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride
16219577
288.82 (270.80 Anhydrous)
Crystalline Powder
75°C to 79°C
C14H25ClN2O2
5 g
14,5482
Soluble in water (50mg/ml),chloroform,ethanol,and benzene. Insoluble in diethyl ether.
O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C
288.82
CHEBI:60791
98%
Lidocaine hydrochloride monohydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : AN7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only