missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+)-Limonene, 96%, unstabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 179395000

 View more versions of this product

Catalog No. AC179395000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(+)-Limonene
95.0
-74.0°C
0.8405g/mL
48°C
95% min. (GC)
C10H16
MFCD00062991
2052
0.8405
+106.00 (20.00°C c=1,CH3OH)
d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene
XMGQYMWWDOXHJM-JTQLQIEISA-N
(4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
440917
CHEBI:15382
96%
5989-27-5,10191-41-0
100.0
Colorless to Yellow
175.4°C to 176.0°C
Authentic
Glass Bottle
1.4700 to 1.4740
500 mL
05, 133
14, 5493
+ 106.00
Solubility in water: insoluble. Other solubilities: miscible with alcohol
CC(=C)[C@@H]1CCC(C)=CC1
136.24
0.846 mPa.s (25°C)
136.24
Liquid

Chemical Identifiers

5989-27-5
136.24
XMGQYMWWDOXHJM-JTQLQIEISA-N
440917
CC(=C)[C@@H]1CCC(C)=CC1
C10H16
MFCD00062991
d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene
CHEBI:15382
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

EINECSNumber : 227-813-5

RTECSNumber : GW6360000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only