missing translation for 'onlineSavingsMsg'
Learn More

Linalool, 97%

CAS: 78-70-6 | C10H18O | 154.25 g/mol

$117.63 - $750.38

Chemical Identifiers

CAS 78-70-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00008906
InChI Key CDOSHBSSFJOMGT-UHFFFAOYSA-N
Synonym linalool, linalol, 3,7-dimethyl-1,6-octadien-3-ol, allo-ocimenol, beta-linalool, linalyl alcohol, 1,6-octadien-3-ol, 3,7-dimethyl, +--linalool, p-linalool, linanool
PubChem CID 6549
ChEBI CHEBI:17580
IUPAC Name 3,7-dimethylocta-1,6-dien-3-ol
SMILES CC(=CCCC(C)(C=C)O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC125151000
SDS View Documents
Thermo Scientific Chemicals
125151000
100 mL Glass bottle
Each for $117.63
Only null left
 
AC125155000
SDS View Documents
Thermo Scientific Chemicals
125155000
500 mL Glass bottle
Each for $259.80
Only null left
 
AC125150025
SDS View Documents
Thermo Scientific Chemicals
125150025
2.5 L Glass bottle
Each for $750.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

78-70-6
154.25
CDOSHBSSFJOMGT-UHFFFAOYSA-N
6549
3,7-dimethylocta-1,6-dien-3-ol
C10H18O
MFCD00008906
linalool, linalol, 3,7-dimethyl-1,6-octadien-3-ol, allo-ocimenol, beta-linalool, linalyl alcohol, 1,6-octadien-3-ol, 3,7-dimethyl, +--linalool, p-linalool, linanool
CHEBI:17580
CC(=CCCC(C)(C=C)O)C

Specifications

78-70-6
0.8610g/mL
75°C
96% min. (GC)
C10H18O
(CH3)2C=CHCH2CH2C(CH3)(OH)CH=CH2
100 mL
0.861
-2° to +2° (20°C, 589nm) (neat)
Solubility in water: 1.45g/L (25°C). Other solubilities: miscible with alcohol and ether
CC(=CCCC(C)(C=C)O)C
154.25
4.4 mPa.s (25°C)
154.25
Liquid
Colorless to Yellow
199.0°C
Authentic
Glass bottle
1.4600 to 1.4640
MFCD00008906
01, 462
15, 5550
linalool, linalol, 3,7-dimethyl-1,6-octadien-3-ol, allo-ocimenol, beta-linalool, linalyl alcohol, 1,6-octadien-3-ol, 3,7-dimethyl, +--linalool, p-linalool, linanool
CDOSHBSSFJOMGT-UHFFFAOYSA-N
3,7-dimethylocta-1,6-dien-3-ol
6549
CHEBI:17580
97%
Linalool, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice

GHS Signal Word: Warning

EINECSNumber : 201-134-4

RTECSNumber : RG5775000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only