Learn More
Thermo Scientific™ Lindberg/Blue M™ Vacuum Ovens
Benefit from a digital control, built-in over-temperature protection and a flexible vacuum/purge/release system for a range of applications with these ovens.
$11600.62 - $18329.76
Specifications
| Chamber Material | Corrosion-resistant Stainless Steel |
|---|---|
| Controller Type | Digital Microprocessor Control |
| Temperature (Metric) | 6°C to 260°C |
| Type | Digital, OTP |
| Display | LED |
Alert:
Ovens are not suitable for use with hazardous vapor. For use with nonflammable, noncorrosive inert gases only.
| Catalog Number | Mfr. No. | Capacity (Metric) | Electrical Requirements | Material | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Electrical Requirements | Material | Price | Quantity | |||
|
13-258-13B
|
Thermo Scientific™
VO1218A1 |
42.5 L | 120 V 50/60 Hz | Painted Steel |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
13-258-13
|
Thermo Scientific™
VO914A1 |
18.6 L | 120 V 50/60 Hz | Painted Steel |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Benefit from a digital control, built-in over-temperature protection and a flexible vacuum/purge/release system with Thermo Scientific™ Lindberg/Blue M™ Vacuum Ovens. These ovens work well with a range of applications including drying, curing, outgassing, aging, process control and other applications requiring elevated temperature in reduced atmospheres or vacuum/purge with nonflammable and inert atmospheres.
Single-Setpoint Digital Microprocessor Control
- Maximum temperature: 260°C (500°F) with nominal uniformity of ±3.5% of setpoint
- Simultaneous LED display of setpoint and actual temperature with push-button view; resolution to within 1°C
- Keypad data entry and pull-out control module for easy service
- Independent overtemperature safety system factory set with reset pushbutton
Vacuum System
- 1 in.(2.54cm) manifold, chamber rear, exterior, connects to pump or in-house vacuum source
- With inert gas injection valve, fresh air inlet, vacuum exhaust port, controllable vacuum release vent port
- 1000 microns (1 torr) capacity
- Vacuum gauge, vacuum, gas and inlet valves located on front
Construction
- Corrosion-resistant stainless-steel cabinet includes three aluminum shelves with painted or stainless-steel finish
- Rear-mounted vacuum manifold connects to pump (not included) or in-house vacuum source
- Low-watt heating element extends oven life
- Heavy-gauge steel, dual-wall construction with fiberglass insulation for minimum heat loss in walls and door
- High-temperature silicone gasket is specially molded for a positive seal
- Magnetic latch for positive door seal (4.5 cu. ft. capacity models include mechanical latches)
- Viewing windows: 0.5 in.(1.3cm) thick in two smaller capacity models or 0.75 in.(1.9cm) thick in 4.5 cu. ft. model; permit observation of work in progress without loss of heat
- Stainless-steel exterior models come with double door—a solid outer door over inner window door
Includes
Power cord and aluminum shelves (number of shelves based on model; see ordering table)
Warranty: 1 year warranty
Warranty
1 year (parts and labor)
Specifications
| Corrosion-resistant Stainless Steel | |
| 6°C to 260°C | |
| LED | |
| Overtemperature Safety System | |
| ±3.5°C | |
| 50/60 Hz |
| Digital Microprocessor Control | |
| Digital, OTP | |
| Inert gas port, vacuum port and vent port | |
| ±1°C | |
| 1 x 10-2 torr (10 microns) |