missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Linoleic acid, 99%
CAS: 60-33-3 | C18H32O2 | 280.45 g/mol
$119.50 - $683.80
Chemical Identifiers
| CAS | 60-33-3 |
|---|---|
| Molecular Formula | C18H32O2 |
| Molecular Weight (g/mol) | 280.45 |
| InChI Key | OYHQOLUKZRVURQ-HZJYTTRNSA-N |
| Synonym | linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid |
| PubChem CID | 5280450 |
| ChEBI | CHEBI:17351 |
| IUPAC Name | (9Z,12Z)-octadeca-9,12-dienoic acid |
| SMILES | CCCCCC=CCC=CCCCCCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC215040050
|
Thermo Scientific Chemicals
215040050 |
5 mL | Glass bottle |
Each for $119.50
|
|
||||
|
AC215040250
|
Thermo Scientific Chemicals
215040250 |
25 mL | Glass bottle |
Each for $343.04
|
|
||||
|
AC215041000
|
Thermo Scientific Chemicals
215041000 |
100 mL | Glass bottle |
Each for $683.80
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 60-33-3 | |
| 280.45 | |
| linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid | |
| CHEBI:17351 | |
| CCCCCC=CCC=CCCCCCCCC(=O)O |
| C18H32O2 | |
| OYHQOLUKZRVURQ-HZJYTTRNSA-N | |
| 5280450 | |
| (9Z,12Z)-octadeca-9,12-dienoic acid |
Specifications
| 60-33-3 | |
| Colorless to Yellow | |
| 229.0°C to 230.0°C (16.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4687 to 1.4707 | |
| 5 mL | |
| 0.9 | |
| linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid | |
| OYHQOLUKZRVURQ-HZJYTTRNSA-N | |
| (9Z,12Z)-octadeca-9,12-dienoic acid | |
| 5280450 | |
| 280.45 | |
| Liquid |
| -5.0°C | |
| 0.9000g/mL | |
| >112°C | |
| 98.5% min. (GC) | |
| C18H32O2 | |
| CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H | |
| 02,IV,1754 | |
| 15,556 | |
| Solubility in water: insoluble. Other solubilities: freely soluble in diethyl ether,miscible with dimethylformamide,soluble in ethanol,petroleum ether | |
| CCCCCC=CCC=CCCCCCCCC(=O)O | |
| 280.45 | |
| CHEBI:17351 | |
| 99% | |
| Linoleic acid |
Safety and Handling
Warning
EINECSNumber : 200-470-9
RUO – Research Use Only