missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Lipofectamine™ 3000 Transfection Reagent DFS Item

Lipofectamine™ 3000 Transfection Reagent leverages our most advanced lipid nanoparticle technology to provide superior transfection performance with improved application outcomes and reproducible results.

$120.00 - $10310.00

Specifications

Classification Animal Origin-Free
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR)
Transfection Technique Lipid-based Transfection
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L3000001 DFS Item
View Documents
Invitrogen™
L3000001
0.1 mL
Each for $120.00
 
L3000008 DFS Item
View Documents
Invitrogen™
L3000008
0.75 mL
Each for $754.00
 
L3000015 DFS Item
View Documents
Invitrogen™
L3000015
1.5 mL
Each for $1,200.00
 
L3000075 DFS Item
View Documents
Invitrogen™
L3000075
5 x 1.5 mL
Each for $5,780.00
 
L3000150 DFS Item
View Documents
Invitrogen™
L3000150
15 mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Lipofectamine™ 3000 Transfection Reagent leverages our most advanced lipid nanoparticle technology to provide superior transfection performance with improved application outcomes and reproducible results.

This reagent delivers superior transfection efficiency and improved cell viability for the widest range of hard-to-transfect and common cells (e.g., HEK293, HeLa).

Successfully transfect the widest variety of biologically relevant cell types with a reagent that offers:
  • Superior performance—our highest efficiency reagent for difficult-to-transfect cells
  • Improved cell viability—gentle on your cells, with low toxicity
  • Versatile—one reagent for DNA, RNA, and co-transfection

Efficiently Transfect Difficult-to-Transfect Cells
Lipofectamine 3000 reagent is designed to efficiently transfect difficult-to-transfect cells, yielding superior transfection performance across the broadest array of cell types. Successfully transfected cell types include:

Fibroblast (3T3, COS-7)
Myoblast (C2C12, L6 CRL-1458)
Hepatocyte (HepG2, HuH7)
Erythroleukemic cell (K562)
Breast carcinoma (MCF7, Hs578T)
Prostate carcinoma (LNCap)
Lung carcinoma (A549, NCI-H460)
Osteosarcoma (U-2 OS, Saos-2)
Colon carcinoma (Caco2, SW480)
Pancreatic carcinoma (PANC-1)
Skin melanoma (SK-MEL-28)

Transfect Common Cells Efficiently and Cost-Effectively
For common cell types, Lipofectamine 3000 reagent is more cost-effective than other reagents due to its high concentration and low usage amount. The 1.5mL size provides up to 1500 transfection reactions (in 24-well plates).

Achieve Superior Transfection Efficiency While Maintaining Low Toxicity
Lipofectamine 3000 reagent maintains a high transfection efficiency within a robust dynamic range of lipid doses for quick and easy optimization. A low toxicity lipid dose (0.75μL for 24-well plates) is suggested for applications requiring minimal disruption of the cells.

Enable High Transfection Efficiency in Novel Genome Editing Applications
Lipofectamine 3000 reagent is suitable for novel genome editing applications. It increases the likelihood of successful cleavage and recombination with TALENs or CRISPRs through highly efficient transfection, ultimately maximizing the efficiency of genetic modifications and simplifying the downstream processes.
Specifications

Specifications

Animal Origin-Free
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Lipid-based Transfection
Sustainable packaging
Transfection Reagent
Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR)
Transfection
Lipofectamine™
Yes
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.