Invitrogen™ Lipofectamine™ RNAiMAX Transfection Reagent DFS Item
Provides highest transfection efficiencies on widest variety of cell types for siRNA-mediated gene knockdown experiments
Supplier:  Invitrogen™ 13778500
 View more versions of this product
Catalog No. 13-778-500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Lipofectamine™ RNAiMAX Transfection Reagent provides the highest transfection efficiencies on the widest variety of cell types for siRNA-mediated gene knockdown experiments. Lipofectamine™ RNAiMAX is a proprietary RNAi-specific cationic lipid formulation designed specifically for the delivery of siRNA and miRNA into all cell types.
With Lipofectamine™ RNAiMAX Transfection Reagent you will get:
  • Superior transfection efficiency requiring lower RNAi concentrations, leading to more effective gene knockdown with minimal nonspecific effects
  • Easy optimization due to minimal cytotoxicity across a 10-fold concentration range of transfection reagent
  • Superior transfection efficiencies for miRNA antagonists and mimics
  • Compatibility with a broad range of cell types, providing the most versatile approach to all of your gene silencing experiments
  • A simple and rapid protocol for consistent and reproducible results
    High knockdown in a wide range of cells
    Lipofectamine™ RNAiMAX Transfection Reagent transfects a wide range of cell types (see figure). For gene silencing, Lipofectamine™ RNAiMAX Transfection Reagent's high-efficiency transfections lead to the high levels of gene knockdown needed to achieve convincing results.
    Simple, high-throughput–ready transfections
    Simply mix Lipofectamine™ RNAiMAX Transfection Reagent with siRNA, add to your cells, incubate, and measure gene knockdown. The simplicity and speed combined with high transfection efficiency make Lipofectamine™ RNAiMAX Transfection Reagent ideal for high-throughput siRNA transfections. Transfection conditions can be easily established for automated or robotic systems used in such applications.
    Find an optimized Lipofectamine™ RNAiMAX transfection protocol for your cell line.
  • Specifications
    Specifications
    Each unit contains one vial of Lipofectamine™ RNAiMAX™ Reagent (15 ml). Store at 2–8°C.
    6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
    Lipid-based Transfection
    Sustainable packaging
    Lipofectamine™
    15 mL
    Approved for shipment at Room Temperature or on Wet Ice
    Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
    siRNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR), RNAi, miRNA
    Transfection
    High-throughput Compatible
    Transfection Reagent
    Yes
    Videos
    SDS
    Documents
    Documents
    Promotions
    Promotions
    For Research Use Only. Not for use in diagnostic procedures.