missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Liquefied Phenol, USP, 89%, Spectrum™ Chemical

PH125, 108-95-2, C6H6O

$976.00 - $35007.57

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
IUPAC Name phenol
SMILES OC1=CC=CC=C1
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18603911
View Documents
Spectrum Chemical Mfg Cor
PH125500MLGL
500 mL Poly-Coated Glass Bottle
Each for $976.00
 
18603912
View Documents
Spectrum Chemical Mfg Cor
PH125500MLGLCS6
6 x 500 mL Poly-Coated Glass Bottle
Case of 6 Each for $4,545.34
 
18603908
View Documents
Spectrum Chemical Mfg Cor
PH1254LTPV
4 L Poly-Coated Glass Bottle
Each for $3,311.17
 
18603910
View Documents
Spectrum Chemical Mfg Cor
PH1254LTPVCS4
4 x 4 L Poly-Coated Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18603913
View Documents
Spectrum Chemical Mfg Cor
PH125200LTBL
200 L Poly-Coated Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Liquefied Phenol, USP is commonly used for permanent treatment of ingrown toe and finger nails, a procedure known as a chemical matrixectomy. All Spectrum Chemical USP products USP are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

108-95-2
94.11
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
OC1=CC=CC=C1
C6H6O
MFCD00002143
phenol

Specifications

108-95-2,7732-18-5
Poly-Coated Glass Bottle
MFCD00002143
UN2821
OC1=CC=CC=C1
94.11
USP
89%
C6H6O
500 mL
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
phenol
89%
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'