missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Science First™ Liquid Conductivity Indicator
Determine the conductivity of different liquids or concentrations.
Supplier: Science First™ S43478BLB
Exige : Light bulb, 7.5w
Description
- Qualitative device causes a 7.5w light bulb to glow when water conducts electricity
- Cord on the side cannot contact water, eliminating shock hazards
- Plastic molded body with recessed electrical connections and electrodes
- Attaches to standard ring stand (up to 0.5 in. dia. [12.7mm]) with thumbscrew clamp (sold separately)
- Comes with illustrated instructions
- Weight: 3.88 oz. (0.11kg)
Operates on 110V, AC only
Spécifications
| Determine the conductivity of liquids or concentrations | |
| Replacement Bulb | |
| 7.5 w |