Agilent BioTek Liquid Handling Control Software, USB Delivery
Liquid Handling Control Software provides a powerful yet flexible interface for use with washers, dispensers and combination liquid handling systems. This USB delivery is to be purchased in conjunction with LHC3 or LHC3SECURE.
Supplier: Agilent BioTek LHC3USB
Description
- LHC software makes it easy to program, even the most complex protocols with a choice of deployment
- Protocols can be run directly from the computer or downloaded to the instrument for convenient onboard recall and operation
- Full instrument control
- For use with 405 TS Washer, 405 LS Washer, ELx405 Select Deep Well Washer, 50 TS Washer, MultiFlo Dispenser, MultiFlo FX Dispenser, EL406 Washer Dispenser, 406 FX Washer Dispenser
- Batch processing with connected BioStack Microplate Stacker or BioSpa 8 Automated Incubator
- Seamless robotic integration to commercially available robotic systems
Specifications
USB Flash Drive Delivery | |
Agilent BioTek Microplate Washers, Dispensers, and Liquid Handling Systems | |
Liquid Handling Control Software |