missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium aluminum Hydride, 2.4M Solution in THF, AcroSeal™, Thermo Scientific Chemicals
$163.59 - $531.60
Specifications
Chemical Name or Material | Lithium Aluminum hydride |
---|---|
Color | Yellow |
Name Note | 2.4M Solution in THF |
CAS | 16853-85-3,109-99-9 |
Molecular Formula | AlH4Li |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC377321000
|
Thermo Scientific Chemicals
377321000 |
100 mL | AcroSeal™ Glass bottle |
Each for $163.59
|
|
||||
AC377328000
|
Thermo Scientific Chemicals
377328000 |
800 mL | AcroSeal™ Glass Bottle |
Each for $531.60
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Lithium Aluminum hydride | |
2.4M Solution in THF | |
AlH4Li | |
MFCD00011075 | |
9.5 to 10.5% (as LiAlH4) | |
15, 344 | |
Solubility in water: vigorous reaction. | |
[Li+].[AlH4-] | |
37.95 | |
37.95 | |
0.9 | |
Viscous Liquid |
Yellow | |
16853-85-3,109-99-9 | |
LiAlH4 | |
1411 | |
0.9000g/mL | |
lithium aluminum hydride, lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride | |
OCZDCIYGECBNKL-UHFFFAOYSA-N | |
lithium(1+) alumanuide | |
21226445 | |
9.2 to 10.5% (as LiAlH4) | |
−17°C |
RUO – Research Use Only