missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Bis(fluorosulfonyl)imide 98.0+%, TCI America™

Supplier:  TCI America L02815G

 View more versions of this product

Catalog No. L02815G


Explore available promotions

Description

Description

Specifications

Specifications

Lithium Bis(fluorosulfonyl)imide
133°C
F2LiNO4S2
5 g
LiFSI
[Li+].FS(=O)(=O)[N-]S(F)(=O)=O
187.06
187.06
Crystalline Powder
171611-11-3
White-Yellow
MFCD28126786
1759
VDVLPSWVDYJFRW-UHFFFAOYSA-N
lithium(1+) bis(fluorosulfonyl)azanide
86277430
≥98.0% (T)

Chemical Identifiers

171611-11-3
187.06
VDVLPSWVDYJFRW-UHFFFAOYSA-N
86277430
[Li+].FS(=O)(=O)[N-]S(F)(=O)=O
F2LiNO4S2
MFCD28126786
LiFSI
lithium(1+) bis(fluorosulfonyl)azanide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (1)-1247

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions