missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium Bis(pentafluoroethanesulfonyl)imide 98.0+%, TCI America™
Supplier: TCI America L02671G
Specifications
Lithium Bis(pentafluoroethanesulfonyl)imide | |
328°C | |
C4F10LiNO4S2 | |
1 g | |
Bis(pentafluoroethanesulfonyl)imide Lithium Salt | |
[Li+].FC(F)(F)C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)C(F)(F)F | |
387.09 | |
387.09 | |
Crystalline Powder |
132843-44-8 | |
White | |
MFCD22374096 | |
2923 | |
ACFSQHQYDZIPRL-UHFFFAOYSA-N | |
lithium(1+) 1,1,1,2,2-pentafluoro-2-{[(1,1,2,2,2-pentafluoroethanesulfonyl)azanidyl]sulfonyl}ethane | |
12096715 | |
≥98.0% (T) |
Chemical Identifiers
132843-44-8 | |
387.09 | |
ACFSQHQYDZIPRL-UHFFFAOYSA-N | |
12096715 | |
[Li+].FC(F)(F)C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)C(F)(F)F |
C4F10LiNO4S2 | |
MFCD22374096 | |
Bis(pentafluoroethanesulfonyl)imide Lithium Salt | |
lithium(1+) 1,1,1,2,2-pentafluoro-2-{[(1,1,2,2,2-pentafluoroethanesulfonyl)azanidyl]sulfonyl}ethane |
Safety and Handling
EINECSNumber : (2)-3839
TSCA : Yes