missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium bis(trimethylsilyl)amide, 1M solution in THF/Ethylbenzene, AcroSeal™
CAS: 4039-32-1 | C6H18LiNSi2 | 167.33 g/mol
$122.60 - $122.60
Specifications
| Chemical Name or Material | Lithium bis(trimethylsilyl)amide |
|---|---|
| Name Note | 1M Solution in THF/Ethylbenzene |
| CAS | 4039-32-1,109-99-9,100-41-4 |
| Color | Yellow to Brown |
| Density | 0.8900g/mL |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC347701000
|
Thermo Scientific Chemicals
347701000 |
100 mL | AcroSeal™ Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Lithium bis(trimethylsilyl)amide | |
| 4039-32-1,109-99-9,100-41-4 | |
| 0.8900g/mL | |
| C6H18LiNSi2 | |
| 04,296; 05,393; 07,197; 12,280; 13,165; 14,194 | |
| lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide | |
| YNESATAKKCNGOF-UHFFFAOYSA-N | |
| lithium;bis(trimethylsilyl)azanide | |
| 2733832 | |
| 18 to 22% active base (as LiNSi) |
| 1M Solution in THF/Ethylbenzene | |
| Yellow to Brown | |
| −21°C | |
| MFCD00008261 | |
| 0.89 | |
| Solubility in water: decomposes | |
| [Li+].C[Si](C)(C)[N-][Si](C)(C)C | |
| 167.33 | |
| 167.33 | |
| Turbid Solution |
RUO – Research Use Only