Learn More
Lithium carbonate, 99+%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 413261000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium carbonate | |
554-13-2 | |
Fine Crystalline Powder | |
99+% | |
Li2CO3 | |
01,606; 03,183; 04,192; 05,396 | |
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
XGZVUEUWXADBQD-UHFFFAOYSA-L | |
dilithium(1+) carbonate | |
11125 | |
73.88 | |
ACS Reagent | |
2.11g/mL |
720.0°C | |
White | |
100 g | |
CLi2O3 | |
MFCD00011084 | |
15,5583 | |
Solubility in water: 13g/L (20°C). Other solubilities: soluble in diluted acids,practically insoluble in alcohol | |
[Li+].[Li+].[O-]C([O-])=O | |
73.89 | |
CHEBI:6504 | |
≥99% | |
Glass bottle |
Chemical Identifiers
554-13-2 | |
73.89 | |
XGZVUEUWXADBQD-UHFFFAOYSA-L | |
11125 | |
dilithium(1+) carbonate |
CLi2O3 | |
MFCD00011084 | |
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
CHEBI:6504 | |
[Li+].[Li+].[O-]C([O-])=O |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Warning
EINECSNumber : 209-062-5
RUO – Research Use Only