missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium chloride, 99%, For Analysis, Anhydrous, Thermo Scientific Chemicals

Lithium Chloride, 99%, ClLi, CAS Number-7447-41-8, chlorure de lithium, chlorku litu polish, chlorku litu, lithium chloride licl, lithium chloride, licl, luthium chloride, lithiumchloride, lithiumchlorid, chlorolithium, 500g, carbonate (as Li2CO3): < 0.03% (typically), 98.5% min. (Argentometry)

$103.36 - $3142.93

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC199881000
SDS View Documents
Thermo Scientific Chemicals
199881000
100 g Plastic bottle
Each for $103.36
 
AC199885000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
199885000
500 g Plastic bottle
Each for $256.67
 
AC199880020
SDS View Documents
Thermo Scientific Chemicals
199880020
2 kg Plastic bottle
Each for $891.57
 
AC199880100
SDS View Documents
Thermo Scientific Chemicals
199880100
10 kg Plastic drum
Each for $3,142.93
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7447-41-8
42.39
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
433294
lithium(1+) chloride
ClLi
MFCD00011078
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
CHEBI:48607
[Li+].[Cl-]

Specifications

1382.0°C
7447-41-8
Crystalline Granules
98.5% min. (Argentometry)
LiCl
01,609; 02,246; 04,298; 05,677; 12,277; 16,194
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium(1+) chloride
433294
42.38
Analytical
Plastic bottle
2.0600g/mL
605.0°C
White
100 g
ClLi
MFCD00011078
15,5584
Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols, ether, pyridine,, nitrobenzene and acetone
[Li+].[Cl-]
42.39
CHEBI:48607
99%
0.5% max. (400°C, 4 hrs)
2.06
Lithium chloride, 99%
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse

GHS Signal Word: Warning

EINECSNumber : 231-212-3

RTECSNumber : OJ5950000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only