Learn More
Lithium chloride, 99%, extra pure, Thermo Scientific Chemicals
$2393.18 - $2393.18
Chemical Identifiers
CAS | 7447-41-8 |
---|---|
Molecular Formula | ClLi |
Molecular Weight (g/mol) | 42.39 |
MDL Number | MFCD00011078 |
InChI Key | KWGKDLIKAYFUFQ-UHFFFAOYSA-M |
Synonym | lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride |
PubChem CID | 433294 |
ChEBI | CHEBI:48607 |
IUPAC Name | lithium(1+) chloride |
SMILES | [Li+].[Cl-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC193370050
|
Thermo Scientific Chemicals
193370050 |
5 kg | Plastic bottle |
Each for $2,393.18
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7447-41-8 | |
42.39 | |
KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
433294 | |
lithium(1+) chloride |
ClLi | |
MFCD00011078 | |
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride | |
CHEBI:48607 | |
[Li+].[Cl-] |
Specifications
1382.0°C | |
7447-41-8 | |
98.5% min. (Argentometry) | |
LiCl | |
01,609; 02,246; 04,298; 05,677; 12,277; 16,194 | |
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride | |
KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
lithium(1+) chloride | |
433294 | |
42.38 | |
Extra Pure | |
2.06 | |
Lithium chloride |
605.0°C | |
5 kg | |
ClLi | |
MFCD00011078 | |
15,5584 | |
Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols, ether, pyridine,, nitrobenzene and acetone | |
[Li+].[Cl-] | |
42.39 | |
CHEBI:48607 | |
99% | |
Plastic bottle | |
2.0600g/mL |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse
GHS Signal Word: Warning
EINECSNumber : 231-212-3
RUO – Research Use Only