missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride-potassium chloride

CAS: 65567-96-6 | Cl2KLi | 116.938 g/mol

$236.55 - $896.78

Chemical Identifiers

CAS 65567-96-6
Molecular Formula Cl2KLi
Molecular Weight (g/mol) 116.938
MDL Number MFCD00144623
InChI Key HCQWRNRRURULEY-UHFFFAOYSA-L
Synonym lithium chloride-potassium chloride, acmc-20akmw, lithium potassium chloride, lithium potassium chloride 1/1/2, lithium chloride/potassium chloride eutectic, lithium 1+ ion potassium dichloride, lithium chloride/potassium chloride eutectic, anhydrous, beads trace metals basis
PubChem CID 16213218
IUPAC Name lithium;potassium;dichloride
SMILES [Li+].[Cl-].[Cl-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1820618
View Documents
Thermo Scientific Chemicals
01820618
50 g N/A
Only null left
 
AA1820630
View Documents
Thermo Scientific Chemicals
01820630
250 g N/A
Only null left
 
Description

Description

Lithium chloride-potassium chloride is used in separators for mixed transition-metal oxide cathodes in thermal batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

65567-96-6
116.938
HCQWRNRRURULEY-UHFFFAOYSA-L
16213218
[Li+].[Cl-].[Cl-].[K+]
Cl2KLi
MFCD00144623
lithium chloride-potassium chloride, acmc-20akmw, lithium potassium chloride, lithium potassium chloride 1/1/2, lithium chloride/potassium chloride eutectic, lithium 1+ ion potassium dichloride, lithium chloride/potassium chloride eutectic, anhydrous, beads trace metals basis
lithium;potassium;dichloride

Specifications

357°C
65567-96-6
50 g
MFCD00144623
Soluble in water.
[Li+].[Cl-].[Cl-].[K+]
116.938
Odorless
2.02
White
Crystals
Cl2KLi
lithium chloride-potassium chloride, acmc-20akmw, lithium potassium chloride, lithium potassium chloride 1/1/2, lithium chloride/potassium chloride eutectic, lithium 1+ ion potassium dichloride, lithium chloride/potassium chloride eutectic, anhydrous, beads trace metals basis
HCQWRNRRURULEY-UHFFFAOYSA-L
lithium;potassium;dichloride
16213218
Hygroscopic
Lithium chloride-potassium chloride
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only