Learn More
Lithium diisopropylamide, 2M sol. in THF/n-heptane/ethylbenzene, AcroSeal™, Thermo Scientific Chemicals
$117.15 - $433.75
Chemical Identifiers
CAS | 4111-54-0 |
---|---|
Molecular Formula | C6H14LiN |
Molecular Weight (g/mol) | 107.125 |
MDL Number | MFCD00064449 |
InChI Key | ZCSHNCUQKCANBX-UHFFFAOYSA-N |
Synonym | lithium diisopropylamide, unii-ol028kiw1i, lithium di-isopropylamide, lithium diisopropyl amide, 2-propanamine, n-1-methylethyl-, lithium salt, ol028kiw1i, lithium di propan-2-yl azanide, 2-propanamine, n-1-methylethyl-, lithium salt 1:1, lithium n,n-diisopropylamide, ipr2nli |
PubChem CID | 2724682 |
IUPAC Name | lithium;di(propan-2-yl)azanide |
SMILES | [Li+].CC(C)[N-]C(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC268831000
|
Thermo Scientific Chemicals
268831000 |
100 mL | AcroSeal™ Glass bottle |
Each for $117.15
|
|
||||
AC268838000
|
Thermo Scientific Chemicals
268838000 |
800 mL | AcroSeal™ Glass Bottle |
Each for $433.75
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4111-54-0 | |
107.125 | |
ZCSHNCUQKCANBX-UHFFFAOYSA-N | |
2724682 | |
[Li+].CC(C)[N-]C(C)C |
C6H14LiN | |
MFCD00064449 | |
lithium diisopropylamide, unii-ol028kiw1i, lithium di-isopropylamide, lithium diisopropyl amide, 2-propanamine, n-1-methylethyl-, lithium salt, ol028kiw1i, lithium di propan-2-yl azanide, 2-propanamine, n-1-methylethyl-, lithium salt 1:1, lithium n,n-diisopropylamide, ipr2nli | |
lithium;di(propan-2-yl)azanide |
Specifications
Brown-Red or Orange to Yellow | |
60.0°C to 136.0°C | |
MFCD00064449 | |
Lithium diisopropylamide: 26.0% (2.0M) | |
0.8100g/mL | |
lithium diisopropylamide, unii-ol028kiw1i, lithium di-isopropylamide, lithium diisopropyl amide, 2-propanamine, n-1-methylethyl-, lithium salt, ol028kiw1i, lithium di propan-2-yl azanide, 2-propanamine, n-1-methylethyl-, lithium salt 1:1, lithium n,n-diisopropylamide, ipr2nli | |
ZCSHNCUQKCANBX-UHFFFAOYSA-N | |
lithium;di(propan-2-yl)azanide | |
2724682 | |
24.6 to 30.0% (Lithium diisopropylamide) | |
0.81 | |
2°C | |
Lithium diisopropylamide, 2M Solution in THF/n-Heptane |
4111-54-0,142-82-5,109-99-9,100-41-4,108-18-9 | |
C6H14LiN | |
Solvent: THF 59%, n-Heptane: 28%, Ethylbenzene: 13% | |
02,49; 03,184; 04,298; 05,400; 06,334; 07,204; 08,292; 09,280; 10,241; 11,296; 12,277; 14,193; 15,188; 16,15; 17,165 | |
15, 3215 | |
Solubility in water: decomposes. Other solubilities: soluble in thf, n-heptane, ethylbenzene, miscible with liquid aliphatic and aromatic, hydrocarbons, ethers and tertiary amines | |
[Li+].CC(C)[N-]C(C)C | |
107.125 | |
107.12 | |
AcroSeal™ Glass bottle | |
100 mL | |
Solution |
Safety and Handling
GHS P Statement May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. Toxic to aquatic life with long lasting effects. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Danger
EINECSNumber : 223-893-0
TSCA : TSCA
Recommended Storage : Keep the solution at < 10°C at which it is stable
RUO – Research Use Only