missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, 97%, for analysis

CAS: 7789-24-4 | FLi | 25.94 g/mol

$410.52 - $1989.26

Chemical Identifiers

CAS 7789-24-4
Molecular Formula FLi
Molecular Weight (g/mol) 25.94
MDL Number MFCD00011090
InChI Key PQXKHYXIUOZZFA-UHFFFAOYSA-M
Synonym lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PubChem CID 224478
SMILES [Li+].[F-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC218275000
View Documents
Thermo Scientific Chemicals
218275000
500 g Plastic bottle
Each for $410.52
Only null left
 
AC218270025
View Documents
Thermo Scientific Chemicals
218270025
2.5 kg Plastic bottle
Each for $1,989.26
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-24-4
25.94
PQXKHYXIUOZZFA-UHFFFAOYSA-M
224478
FLi
MFCD00011090
lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
[Li+].[F-]

Specifications

1681.0°C
7789-24-4
Powder
96% min
LiF
01,933
lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium(1+) fluoride
224478
97%
0.4% (200°C) (typical)
2.63
Lithium fluoride
845.0°C
White
500 g
FLi
MFCD00011090
15,5587
Solubility in water: 0.29 g/100mL (20°C). Other solubilities: soluble in acids, insoluble in alcohol
[Li+].[F-]
25.94
25.93
Analytical
Plastic bottle
2.6300g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Contact with acids liberates very toxic gas.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persist

GHS Signal Word: Danger

EINECSNumber : 232-152-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only