missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium formate hydrate, 98%, Thermo Scientific Chemicals

$71.22 - $288.53

Chemical Identifiers

CAS 6108-23-2
Molecular Formula CHLiO2
Molecular Weight (g/mol) 51.96
MDL Number MFCD00050823
InChI Key XKPJKVVZOOEMPK-UHFFFAOYSA-M
Synonym lithium formate monohydrate, unii-zqs5s890b6, formic acid, lithium salt, monohydrate, lithium formate hydrate, monolithium formate monohydrate, lithium formate monohydrate mi, acmc-209mow, lithotab formira hydrate, li.cho2.h2o, lithium formate, reagent
PubChem CID 23702287
IUPAC Name lithium;formate;hydrate
SMILES [Li+].[O-]C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1743218
View Documents
Thermo Scientific Chemicals
A1743218
50 g N/A
 
AAA1743230
View Documents
Thermo Scientific Chemicals
A1743230
250 g N/A
 
Description

Description

It is used in the investigation of signal fading in lithium formate EPR dosimeters using a new sensitive method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the investigation of signal fading in lithium formate EPR dosimeters using a new sensitive method.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

6108-23-2
51.96
XKPJKVVZOOEMPK-UHFFFAOYSA-M
23702287
[Li+].[O-]C=O
CHLiO2
MFCD00050823
lithium formate monohydrate, unii-zqs5s890b6, formic acid, lithium salt, monohydrate, lithium formate hydrate, monolithium formate monohydrate, lithium formate monohydrate mi, acmc-209mow, lithotab formira hydrate, li.cho2.h2o, lithium formate, reagent
lithium;formate;hydrate

Specifications

94°C (decomposition)
50 g
MFCD00050823
Soluble in water.
[Li+].[O-]C=O
51.96
51.96 (Anhydrous)
Mild
6108-23-2
CHLiO2
lithium formate monohydrate, unii-zqs5s890b6, formic acid, lithium salt, monohydrate, lithium formate hydrate, monolithium formate monohydrate, lithium formate monohydrate mi, acmc-209mow, lithotab formira hydrate, li.cho2.h2o, lithium formate, reagent
XKPJKVVZOOEMPK-UHFFFAOYSA-M
lithium;formate;hydrate
23702287
98%
Lithium formate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-133-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only