missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium granules, 1-6mm (0.04-0.2in), 99%, Thermo Scientific Chemicals

 

$241.05 - $900.30

Chemical Identifiers

CAS 7439-93-2
Molecular Formula Li
Molecular Weight (g/mol) 6.94
MDL Number MFCD00134051
InChI Key WHXSMMKQMYFTQS-UHFFFAOYSA-N
Synonym litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
PubChem CID 3028194
ChEBI CHEBI:30145
IUPAC Name lithium
SMILES [Li]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA0073318
View Documents
Thermo Scientific Chemicals
0073318
50 g
Each for $241.05
 
AA0073330
View Documents
Thermo Scientific Chemicals
00073330
250 g
Each for $900.30
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
Specifications

Chemical Identifiers

7439-93-2
6.94
WHXSMMKQMYFTQS-UHFFFAOYSA-N
3028194
lithium
Li
MFCD00134051
litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
CHEBI:30145
[Li]

Specifications

Gray
Gray granules
99% (Metals basis)
MFCD00134051
litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
[Li]
6.94
CHEBI:30145
Under Argon
Lithium
7439-93-2
0.53g/cm3 at 20°C
Li
50 g
WHXSMMKQMYFTQS-UHFFFAOYSA-N
lithium
3028194
Odorless
Granular
Videos
Safety and Handling

Safety and Handling

Keep container dry. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H260-H314

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 231-102-5

RTECSNumber : OJ5540000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only