missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide monohydrate, extra pure, ca. 56% LiOH

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol

Supplier:  Thermo Scientific Chemicals 199540250

 View more versions of this product

Catalog No. AC199540250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

920.0°C
462.0°C
Cream to White or Yellow
25 g
LiOH·H2O
07,208; 12,282
lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
GLXDVVHUTZTUQK-UHFFFAOYSA-M
lithium(1+) hydrate hydroxide
168937
∼56% (LiOH)
Plastic bottle
Lithium hydroxide monohydrate
1310-66-3
Crystalline Powder
H3LiO2
MFCD00149772
14,5534
Solubility in water: 109g/L (20°C). Other solubilities: 107g/L (0°C), 148g/L (100°C), slightly soluble in alcohol
[Li+].O.[OH-]
41.96
41.95
Extra Pure

Chemical Identifiers

1310-66-3
41.96
GLXDVVHUTZTUQK-UHFFFAOYSA-M
168937
H3LiO2
MFCD00149772
lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
[Li+].O.[OH-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only