missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide monohydrate, LiOH 56.5% min

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol

$50.48 - $1447.55

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
SMILES [Li+].O.[OH-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1340918
View Documents
Thermo Scientific Chemicals
01340918
50 g
Each for $50.48
Only null left
 
AA1340936
View Documents
Thermo Scientific Chemicals
01340936
500 g
Each for $436.03
Only null left
 
AA13409A3
View Documents
Thermo Scientific Chemicals
013409A3
2 kg
Each for $1,447.55
Only null left
 
Description

Description

Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

1310-66-3
41.96
GLXDVVHUTZTUQK-UHFFFAOYSA-M
168937
H3LiO2
MFCD00149772
lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
[Li+].O.[OH-]

Specifications

1310-66-3
50 g
H3LiO2
lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
GLXDVVHUTZTUQK-UHFFFAOYSA-M
lithium(1+) hydrate hydroxide
168937
Air sensitive; Hygroscopic
Lithium hydroxide monohydrate
Powder
56.5% min
MFCD00149772
Soluble in water,methanol and ethanol.
[Li+].O.[OH-]
41.96
41.96 (23.95 Anhydrous)
1.51 g/mL
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 215-183-4

RTECSNumber : OJ6307080

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only